Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Aminomethyl-1-Boc-pyrrolidine, 97%

Catalog No. p-7034721
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52419MD 250 mg
AAH5241903 1 g
AAH5241906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52419MD Supplier Thermo Scientific Chemicals Supplier No. H52419MD
Add to Cart

CAS: 259537-92-3 | C10H20N2O2 | 200.282 g/mol

Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259537-92-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03419256
InChI Key SOGXYCNKQQJEED-MRVPVSSYSA-N
Synonym r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine
PubChem CID 1512534
IUPAC Name tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CN

Specifications

Boiling Point 98°C to 110°C (1 mmHg)
Quantity 250 mg
UN Number UN2735
Solubility Information Slightly soluble in water.
Optical Rotation +47° (c=1 in Chloroform)
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (R)-2-Aminomethyl-1-Boc-pyrrolidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.