Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid | 70874-05-4 | 96.2% | C₁₄H₁₉NO₄ | 100 MG
SDP

Catalog No. 5000434609
Encompass_Preferred

This chemical compound is a leucine derivative, identified by CAS number 70874-05-4. It has a molecular formula of C₁₄H₁₉NO₄ and a molecular weight of 265.30, appearing as a colorless to light yellow liquid with an HPLC purity of 96.17%.

  • High purity of 96.17% as determined by HPLC
  • Identified as a leucine derivative
  • Molecular formula C₁₄H₁₉NO₄
  • Molecular weight 265.30
  • Appears as a colorless to light yellow liquid
  • Defined storage guidelines for pure form and in solvent

Catalog No. 50-004-34609 Supplier Medchemexpress LLC Supplier No. HY77630100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This chemical compound is a leucine derivative, identified by CAS number 70874-05-4. It has a molecular formula of C₁₄H₁₉NO₄ and a molecular weight of 265.30, appearing as a colorless to light yellow liquid with an HPLC purity of 96.17%.

  • High purity of 96.17% as determined by HPLC
  • Identified as a leucine derivative
  • Molecular formula C₁₄H₁₉NO₄
  • Molecular weight 265.30
  • Appears as a colorless to light yellow liquid
  • Defined storage guidelines for pure form and in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.