Learn More
Medchemexpress LLC (R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid | 70874-05-4 | MFCD07368370 | 96.17% | C14H19NO4 | 500 MG
SDP
Catalog No. 5000432713
Encompass_Preferred
(R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid is a leucine derivative. It is shipped at room temperature and is suitable for various laboratory applications. This product is available in 100 mg size, with long-term storage recommended at -20°C for up to 3 years as a powder, or 4°C for up to 2 years as a powder.
  • Leucine derivative
  • Suitable for various laboratory applications
  • Available in 100 mg size
  • Long-term storage at -20°C for 3 years (powder)
  • Short-term storage at 4°C for 2 years (powder)
Catalog No. 50-004-32713 Supplier Medchemexpress LLC Supplier No. HY77630500MG
(R)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid is a leucine derivative. It is shipped at room temperature and is suitable for various laboratory applications. This product is available in 100 mg size, with long-term storage recommended at -20°C for up to 3 years as a powder, or 4°C for up to 2 years as a powder.
  • Leucine derivative
  • Suitable for various laboratory applications
  • Available in 100 mg size
  • Long-term storage at -20°C for 3 years (powder)
  • Short-term storage at 4°C for 2 years (powder)