Learn More
American Radiolabeled Chemicals Inc R-2-BROMOPALM AC 1-14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 362350UC
Encompass_Preferred
R-2-Bromopalmitic acid-1-14C 50uCi in ethanol, 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 335.33, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-910