Learn More
American Radiolabeled Chemicals Inc R-2-BROMOPALMITIC AC-9 10-3H
SDP
Catalog No. 5090104938
Encompass_Preferred
R-2-Bromopalmitic acid-9,10-3H 50uCi in ethanol, 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 335.33, Shipped in Dry Ice, Exclusive, (2-Bromohexadecanoic acid)
Catalog No. 50-901-04938 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 127050UC
Add to Cart
Add to Cart
Request a Quote
R-2-Bromopalmitic acid-9,10-3H 50uCi in ethanol, 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 335.33, Shipped in Dry Ice, Exclusive, (2-Bromohexadecanoic acid)