Learn More
eMolecules​ 10009-70-8 | (R)-(+)-2-Bromopropionic acid | Combi-Blocks | MFCD00145205 | 152.975 | C3H5BrO2 | 96.000 | C[C@@H](Br)C(O)=O | 25g | 296393039
Supplier:  eMolecules​ 296393039
Encompass_Preferred
(R)-(+)-2-Bromopropionic acid | Combi-Blocks | 10009-70-8 | MFCD00145205 | 152.975 | C3H5BrO2 | 96.000 | C[C@@H](Br)C(O)=O | 25g | 296393039
Catalog No. NC2038917