Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-(But-3-en-1-yl)oxirane | 137688-20-1 | 99.9% | C6H10O | 250 MG
SDP

Catalog No. 5000358581
Encompass_Preferred

(R)-2-(But-3-en-1-yl)oxirane is a biological molecule appearing as a colorless to light yellow oil. It is stable under recommended storage conditions and intended for research use only.

  • Appears as a colorless to light yellow oil
  • Stable under recommended storage conditions
  • Soluble in DMSO (100 mg/mL)
  • Store at room temperature for 3 years; in solvent at -80°C for 2 years or -20°C for 1 year
  • For research use only

Catalog No. 50-003-58581 Supplier Medchemexpress LLC Supplier No. 250MGHY46575
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-2-(But-3-en-1-yl)oxirane is a biological molecule appearing as a colorless to light yellow oil. It is stable under recommended storage conditions and intended for research use only.

  • Appears as a colorless to light yellow oil
  • Stable under recommended storage conditions
  • Soluble in DMSO (100 mg/mL)
  • Store at room temperature for 3 years; in solvent at -80°C for 2 years or -20°C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.