Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-(-)-2-butanol | 14898-79-4 | MFCD00064280 | 100.0% | 74.12 g/mol | C4H10O | 1 ML
SDP

Catalog No. 5000208536
Encompass_Preferred

(R)-(-)-2-Butanol is the R enantiomer of 2-butanol supplied as a 10 mM solution in DMSO (1 mL vial) and as bulk liquid; it is used as a chemical intermediate and reagent in synthesis and research applications.

  • R enantiomer of 2-butanol.
  • CAS number 14898-79-4.
  • Purity 99.98% as reported by manufacturer.
  • Molecular formula C4H10O; molecular weight 74.12 g/mol.
  • Supplied as 10 mM solution in DMSO (1 mL) and in bulk liquid sizes (1 g, 5 g, 10 g, 50 g).
  • Manufacturer notes DMSO solubility ~100 mg/mL; ultrasonic may be needed and use newly opened DMSO due to hygroscopicity.

Catalog No. 50-002-08536 Supplier Medchemexpress LLC Supplier No. HYW08795210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-(-)-2-Butanol is the R enantiomer of 2-butanol supplied as a 10 mM solution in DMSO (1 mL vial) and as bulk liquid; it is used as a chemical intermediate and reagent in synthesis and research applications.

  • R enantiomer of 2-butanol.
  • CAS number 14898-79-4.
  • Purity 99.98% as reported by manufacturer.
  • Molecular formula C4H10O; molecular weight 74.12 g/mol.
  • Supplied as 10 mM solution in DMSO (1 mL) and in bulk liquid sizes (1 g, 5 g, 10 g, 50 g).
  • Manufacturer notes DMSO solubility ~100 mg/mL; ultrasonic may be needed and use newly opened DMSO due to hygroscopicity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.