Learn More
Medchemexpress LLC (R)-(-)-2-butanol | 14898-79-4 | MFCD00064280 | 100.0% | 74.12 g/mol | C4H10O | 5 G
SDP
Catalog No. 5000208351
Encompass_Preferred
(R)-(-)-2-butanol is the R-enantiomer of sec-butanol supplied as a high-purity research chemical for chiral synthesis, analytical standards, and stereospecific reaction studies. It is a colorless to light yellow liquid with molecular weight 74.12 and CAS 14898-79-4.
  • High purity (99.98% as supplied)
  • Molecular formula C4H10O, molecular weight 74.12 g/mol
  • Available as a 5 G pack for laboratory use
  • Liquid appearance; density 0.807 g/cm3
  • Suitable for chiral synthesis and analytical standards
Catalog No. 50-002-08351 Supplier Medchemexpress LLC Supplier No. HYW0879525G
(R)-(-)-2-butanol is the R-enantiomer of sec-butanol supplied as a high-purity research chemical for chiral synthesis, analytical standards, and stereospecific reaction studies. It is a colorless to light yellow liquid with molecular weight 74.12 and CAS 14898-79-4.
  • High purity (99.98% as supplied)
  • Molecular formula C4H10O, molecular weight 74.12 g/mol
  • Available as a 5 G pack for laboratory use
  • Liquid appearance; density 0.807 g/cm3
  • Suitable for chiral synthesis and analytical standards