Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Butanol, 98+%

Catalog No. aal1398306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1398306 5 g
AAL1398303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1398306 Supplier Thermo Scientific Chemicals Supplier No. L1398306
Only null left
Add to Cart

CAS: 14898-79-4 | C4H10O | 74.123 g/mol

(R)-(-)-2-Butanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-2-Butanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Partly miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-79-4
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00064280
InChI Key BTANRVKWQNVYAZ-SCSAIBSYSA-N
Synonym r---2-butanol, r-2-butanol, --2-butanol, 2r-butan-2-ol, r-butan-2-ol, r---sec-butanol, 2-butanol, 2r, unii-dlh38k423j, r---sec-butyl alcohol
PubChem CID 84682
ChEBI CHEBI:35686
IUPAC Name (2R)-butan-2-ol
SMILES CCC(C)O

Specifications

Density 0.806
Boiling Point 99°C to 100°C
Flash Point 26°C (79°F)
Odor Alcohol-like
Refractive Index 1.398
Quantity 5 g
UN Number UN1120
Beilstein 1718764
Merck Index 14,1541
Solubility Information Partly miscible in water.
Optical Rotation −15° (c=10 in Methanol)
Formula Weight 74.12
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-2-Butanol
Show More Show Less
missing translation for 'healthHazard1' H226-H319-H335-H336
missing translation for 'healthHazard2' GHS H Statement
H226-H319-H335-H336
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber 238-967-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.