Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-formamido-4-methylpentanoic acid | 44978-39-4 | 95.0% | 159.18 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYI0630100MG

Encompass_Preferred

(R)-2-formamido-4-methylpentanoic acid is a leucine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress-related tasks, and preventing exercise-induced muscle damage.

  • Molecular weight of 159.18
  • Chemical formula C7H13NO3
  • Appearance as an off-white to brown solid
  • Purity of 95.0%
  • Soluble in DMSO at 100 mg/mL
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-28748


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.