Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-2-hydroxy-3-(tetradecanoyl-d7-oxy)propyl (2-(trimethylammonio)ethyl) phosphate | 00-00-0 | 95.4% | 474.62 | C22H39D7NO7P | 5 MG
SDP

Catalog No. 5000264282
Encompass_Preferred

LysoPC(14:0/0:0)-d7 is a deuterium-labeled lysophosphatidylcholine used as an isotope-labeled endogenous metabolite standard for analytical and quantitative lipidomics studies. It is provided as a white to off-white solid and is intended for use as an internal standard in mass spectrometry workflows.

  • Deuterium-labeled lyso-phosphatidylcholine for isotope-dilution assays.
  • Molecular formula C22H39D7NO7P and molecular weight 474.62.
  • Purity 95.4% by HPLC as reported on the certificate of analysis.
  • Appearance white to off-white solid suitable for analytical use.
  • Storage: powder -20°C up to 3 years, 4°C up to 2 years; in solvent -80°C up to 6 months.
  • Intended for use as an endogenous metabolite/internal standard in lipidomics and quantitative assays.

Catalog No. 50-002-64282 Supplier Medchemexpress LLC Supplier No. HY113123S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

LysoPC(14:0/0:0)-d7 is a deuterium-labeled lysophosphatidylcholine used as an isotope-labeled endogenous metabolite standard for analytical and quantitative lipidomics studies. It is provided as a white to off-white solid and is intended for use as an internal standard in mass spectrometry workflows.

  • Deuterium-labeled lyso-phosphatidylcholine for isotope-dilution assays.
  • Molecular formula C22H39D7NO7P and molecular weight 474.62.
  • Purity 95.4% by HPLC as reported on the certificate of analysis.
  • Appearance white to off-white solid suitable for analytical use.
  • Storage: powder -20°C up to 3 years, 4°C up to 2 years; in solvent -80°C up to 6 months.
  • Intended for use as an endogenous metabolite/internal standard in lipidomics and quantitative assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.