Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-2-Hydroxy-4-phenylbutyric acid, 97%

Catalog No. AAH5280403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5280403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5280403 Supplier Thermo Scientific Chemicals Supplier No. H5280403

missing translation for 'validMainframeMsg'

CAS: 29678-81-7 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29678-81-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00192219
InChI Key JNJCEALGCZSIGB-SECBINFHSA-N
PubChem CID 2759328
IUPAC Name (2R)-2-hydroxy-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCC(C(=O)O)O

Specifications

Melting Point 114°C to 117°C
Quantity 1 g
Beilstein 3201296
Synonym r-2-hydroxy-4-phenylbutyric acid, r-2-hydroxy-4-phenylbutanoic acid, r---2-hydroxy-4-phenylbutyric acid, 2r-2-hydroxy-4-phenylbutanoic acid, pubchem5687, r-2-hydroxy-4-phenylbutyricacid, 2r-hydroxy-4-phenylbutanoic acid, 2 r-hydroxy-4-phenylbutanoic acid, r-2-hydroxy-4-phenyl-butyric acid
Optical Rotation −9.5° (c=2.8 in Ethanol)
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material (R)-(-)-2-Hydroxy-4-phenylbutyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.