Learn More
Cayman Chemical R-2 MethndamIde 10mg
SDP
Catalog No. 502895796
Encompass_Preferred
A CB analog with a methyl group in the (R) configuration at C-2 of the ethanolamine group; is not a FAAH inhibitor and is nearly as susceptible to amide hydrolysis as AEA itself
Catalog No. 50-289-5796 Supplier Cayman Chemical Supplier No. 9007410 MG
A CB analog with a methyl group in the (R) configuration at C-2 of the ethanolamine group; is not a FAAH inhibitor and is nearly as susceptible to amide hydrolysis as AEA itself