Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Methylpyrrolidine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. M2747200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2747200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2747200MG Supplier TCI America Supplier No. M2747200MG

missing translation for 'validMainframeMsg'

Specifications

CAS 135324-85-5
Melting Point 75°C
Color White-Yellow
Molecular Formula C5H12ClN
Quantity 200 mg
MDL Number MFCD06796610
InChI Key JNEIFWYJFOEKIM-NUBCRITNSA-N
SMILES CC1CCCN1.Cl
IUPAC Name (2R)-2-methylpyrrolidine;hydrochloride
Molecular Weight (g/mol) 121.608
PubChem CID 11147704
Formula Weight 121.61
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-2-Methylpyrrolidine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.