Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 98.0+%, TCI America™
SDP

Catalog No. O04601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04601G Supplier TCI America Supplier No. O04601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133676-09-2
Molecular Formula C28H38O5
Molecular Weight (g/mol) 454.607
MDL Number MFCD21603881
InChI Key PLGPDUBTEHIWRH-JOCHJYFZSA-N
Synonym 4-[4-(Hexyloxy)benzoyloxy]benzoic Acid (R)-2-Octyl Ester
PubChem CID 60123248
IUPAC Name [4-[(2R)-octan-2-yl]oxycarbonylphenyl] 4-hexoxybenzoate
SMILES CCCCCCC(C)OC(=O)C1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)OCCCCCC

Specifications

Melting Point 49°C
Color White
Quantity 1 g
Formula Weight 454.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.