Learn More
Medchemexpress LLC R -2-Phenylmorpholi 250mg
SDP
Catalog No. 5000205735
Encompass_Preferred
(R)-2-Phenylmorpholine is a fragment molecule commonly used in organic synthesis and can be involved in the synthesis of STAT5/6 PROTAC degraders or inhibitors[1]
Catalog No. 50-002-05735 Supplier Medchemexpress LLC Supplier No. 250MGHYW093114
(R)-2-Phenylmorpholine is a fragment molecule commonly used in organic synthesis and can be involved in the synthesis of STAT5/6 PROTAC degraders or inhibitors[1]