Learn More
Electron Microscopy Sciences MohrPro R-201 Developer 1 QT
SDP
Catalog No. 5024958
Encompass_Preferred
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.
Catalog No. 50-249-58 Supplier Electron Microscopy Sciences Supplier No. 7496101
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.