Learn More
Electron Microscopy Sciences MohrPro R-202 Fixer 1 QT
SDP
Catalog No. 5024953
Encompass
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.
Catalog No. 50-249-53 Supplier Electron Microscopy Sciences Supplier No. 7496210
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.