Learn More
Electron Microscopy Sciences MohrPro R-204 Defoamer 0.75 OZ
SDP
Catalog No. 5024957
Encompass_Preferred
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.
Catalog No. 50-249-57 Supplier Electron Microscopy Sciences Supplier No. 7496401
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.