Learn More
Medchemexpress LLC (R)-3-(1H-indol-3-yl)-2-(5-(p-tolylethynyl)thiophene-2-sulfonamido)propanoic acid | 203640-27-1 | MFCD22200275 | 99.8% | 464.56 g/mol | C24H20N2O4S2 | 1 ML
SDP
Catalog No. 5000239761
Encompass_Preferred
S 3304 is a small-molecule matrix metalloproteinase (MMP) inhibitor selective for MMP-2 and MMP-9. It is a D-tryptophan-derived compound that contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC). Reported to be orally active and non-cytotoxic in preclinical studies, it has been used to investigate angiogenesis and metastatic colonization.
  • Selective inhibitor of MMP-2 and MMP-9.
  • Contains an alkyne group for click chemistry (CuAAC).
  • Reported oral activity and low cytotoxicity in preclinical models.
  • High reported purity suitable for research applications.
Catalog No. 50-002-39761 Supplier Medchemexpress LLC Supplier No. HY10699210MM 1ML
S 3304 is a small-molecule matrix metalloproteinase (MMP) inhibitor selective for MMP-2 and MMP-9. It is a D-tryptophan-derived compound that contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC). Reported to be orally active and non-cytotoxic in preclinical studies, it has been used to investigate angiogenesis and metastatic colonization.
  • Selective inhibitor of MMP-2 and MMP-9.
  • Contains an alkyne group for click chemistry (CuAAC).
  • Reported oral activity and low cytotoxicity in preclinical models.
  • High reported purity suitable for research applications.