Learn More
Medchemexpress LLC (R)-3-(2-chlorophenyl)-N-(1-(3-methoxyphenyl)ethyl)propan-1-amine hydrochloride | 177172-49-5 | MFCD16038895 | 99.7% | 340.3 g/mol | C18H23Cl2NO | 50 MG
SDP
Catalog No. 5000245692
Encompass_Preferred
Tecalcet hydrochloride is the hydrochloride salt of a calcimimetic research compound that allosterically and positively modulates the calcium-sensing receptor (CaSR), increasing receptor sensitivity to extracellular calcium. It is provided for research use in solid form or as DMSO solutions for in vitro studies of CaSR signaling and calcium homeostasis.
  • Allosteric positive modulator of the calcium-sensing receptor for CaSR signaling studies.
  • Orally active compound used in pharmacology and cellular assays.
  • High reported purity suitable for research applications.
  • Available as solid and as DMSO solutions for flexible dosing.
  • Stable when stored sealed and protected from moisture; solvent storage at low temperatures recommended.
  • Well characterized chemically with reported molecular weight and structural identifiers.
Catalog No. 50-002-45692 Supplier Medchemexpress LLC Supplier No. HY10167A50MG
Tecalcet hydrochloride is the hydrochloride salt of a calcimimetic research compound that allosterically and positively modulates the calcium-sensing receptor (CaSR), increasing receptor sensitivity to extracellular calcium. It is provided for research use in solid form or as DMSO solutions for in vitro studies of CaSR signaling and calcium homeostasis.
  • Allosteric positive modulator of the calcium-sensing receptor for CaSR signaling studies.
  • Orally active compound used in pharmacology and cellular assays.
  • High reported purity suitable for research applications.
  • Available as solid and as DMSO solutions for flexible dosing.
  • Stable when stored sealed and protected from moisture; solvent storage at low temperatures recommended.
  • Well characterized chemically with reported molecular weight and structural identifiers.