Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol 98.0+%, TCI America™
SDP

Catalog No. D44181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44181G 1 g
D44185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44181G Supplier TCI America Supplier No. D44181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65355-08-0
Molecular Formula C20H20Br2O2
Molecular Weight (g/mol) 452.186
MDL Number MFCD03093977
InChI Key WDRTXCNGVVLRSZ-UHFFFAOYSA-N
PubChem CID 3251420
IUPAC Name 3-bromo-1-(3-bromo-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl)-5,6,7,8-tetrahydronaphthalen-2-ol
SMILES C1CCC2=C(C(=C(C=C2C1)Br)O)C3=C4CCCCC4=CC(=C3O)Br

Specifications

Melting Point 149°C
Quantity 1 g
Synonym (R)-(+)-3,3′C-Dibromo-5,5′C,6,6′C,7,7′C,8,8′C-octahydro-2,2′C-dihydroxy-1,1′C-binaphthyl
Formula Weight 452.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-3,3′-Dibromo-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-bi-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.