Learn More
Strem, An Ascensus Company CAS# 157488-65-8. 1g. (R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine, min. 97% (R)-MONOPHOS. MFCD03426988
SDP
Catalog No. 5090110116
Encompass
CAS# 157488-65-8. 1g. (R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine, min. 97% (R)-MONOPHOS. MFCD03426988. Molecular Weight: 359.36. Molecular Formula: C22H18NO2P. Color/form: white xtl. Strem# 15-1232. http://www.strem.com/catalog/v/15-1232/
Catalog No. 50-901-10116 Supplier Strem, An Ascensus Company Supplier No. 1512321G
CAS# 157488-65-8. 1g. (R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine, min. 97% (R)-MONOPHOS. MFCD03426988. Molecular Weight: 359.36. Molecular Formula: C22H18NO2P. Color/form: white xtl. Strem# 15-1232. http://www.strem.com/catalog/v/15-1232/