Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Amino-1,2-propanediol, 98%

Catalog No. AC397880050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397880050 5 g Glass bottle
AC397880250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397880050 Supplier Thermo Scientific Chemicals Supplier No. 397880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 66211-46-9 | C3H9NO2 | 91.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66211-46-9
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
InChI Key KQIGMPWTAHJUMN-GSVOUGTGSA-N
Synonym r-3-amino-1,2-propanediol, r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propandiol, 2r-3-aminopropane-1,2-diol, r-+-3-amino-1,2-propanediol, r-+-amino-1,2-propanediol, 1-aminoglycerol, 1,2-propanediol, 3-amino-, 2r, pubchem5711, r-3-amino-1,2-propanol
PubChem CID 6994409
IUPAC Name (2R)-3-aminopropane-1,2-diol
SMILES C(C(CO)O)N

Specifications

Melting Point 54°C to 56°C
Boiling Point 163°C (15.0 mmHg)
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2CH2CH(OH)CH2OH
Quantity 5 g
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material (R)-3-Amino-1, 2-propanediol
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.