Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-Amino-3-phenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. A2655200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2655200MG 200 mg
A26551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2655200MG Supplier TCI America Supplier No. A2655200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13921-90-9 | C9H11NO2 | 165.192 g/mol

Chemical Identifiers

CAS 13921-90-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD01076237
InChI Key UJOYFRCOTPUKAK-MRVPVSSYSA-N
Synonym D-beta-Phenylalanine
PubChem CID 686703
ChEBI CHEBI:67172
IUPAC Name (3R)-3-amino-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)N

Specifications

Melting Point 211°C
Color White
Quantity 200 mg
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material (R)-3-Amino-3-phenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.