Learn More
Sigma Aldrich (R)-3-amino-4-methyl-pentanoic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1-amino-4-methylpiperazine has been used:
- In synthesis of pyrazolo-pyrazine, -pyridine derivatives and 2-[(4-methylpiperazin-1-yl)iminomethyl]phenol
- As a solvating and stabilizing agent in the preparation of uniform ag nanoparticles(<10nm)
Specifications
Specifications
| Molecular Formula | C6H13NO2 |
| Linear Formula | C6H13NO2 |
| MDL Number | MFCD01076231 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 131.17 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.