Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R)-3-amino-5-methyl-hexanoic acid

Catalog No. CDS008875 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS008875 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS008875 Supplier Sigma Aldrich Supplier No. CDS008875100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium benzenesulfonate has been used in raman spectroscopic determination of molecular structural features in sulfonated polystyrene resins
  • It was used as electrolyte in formation of polypyrrole coatings with varying surface morphology on stainless steel
  • It was used in the synthesis of 1-butyl-3-propanenitrile imidazolium benzenesulfonate

Specifications

Molecular Formula C7H15NO2
Linear Formula C7H15NO2
MDL Number MFCD01076255
Quantity 25 g
Molecular Weight (g/mol) 145.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.