Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-Benzyloxy-1,2-propanediol 98.0+%, TCI America™
SDP

Catalog No. B2141100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2141100MG 100 mg
B21411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2141100MG Supplier TCI America Supplier No. B2141100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56552-80-8
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00067260
InChI Key LWCIBYRXSHRIAP-SNVBAGLBSA-N
Synonym (R)-(+)-Glycerol alpha-Benzyl Ether
PubChem CID 2724795
IUPAC Name (2R)-3-phenylmethoxypropane-1,2-diol
SMILES C1=CC=C(C=C1)COCC(CO)O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-3-Benzyloxy-1,2-propanediol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.