Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-(Boc-amino)-3-(3-hydroxyphenyl)propionic acid, 95%, Thermo Scientific™

Catalog No. AAH52109MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52109MD 250 mg
AAH5210903 1 g
AAH5210906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52109MD Supplier Thermo Scientific Chemicals Supplier No. H52109MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500788-89-6
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.308
MDL Number MFCD03427940
InChI Key KLIMLFKCLBBZKH-LLVKDONJSA-N
Synonym boc-r-3-amino-3-3-hydroxy-phenyl-propionic acid, r-3-boc-amino-3-3-hydroxyphenyl propionic acid, r-3-tert-butoxycarbonyl amino-3-3-hydroxyphenyl propanoic acid, 3r-3-tert-butoxycarbonyl amino-3-3-hydroxyphenyl propanoic acid, benzenepropanoic acid, b-1,1-dimethylethoxy carbonyl amino-3-hydroxy-, br, pubchem24049, pubchem24051, boc-d-beta-m-tyr-oh, boc-d-?-phe 3-oh-oh, boc-d-beta-phe 3-oh-oh
PubChem CID 2761766
IUPAC Name (3R)-3-(3-hydroxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)O

Specifications

Quantity 250 mg
Formula Weight 281.31
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-hydroxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.