Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, 97%, Thermo Scientific™

Catalog No. AAH5200103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5200103 1 g
AAH52001MD 250 mg
AAH5200106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5200103 Supplier Thermo Scientific Chemicals Supplier No. H5200103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, petrochemicals, and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500788-86-3
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD03427937
InChI Key DNXONWHJJNOKEU-GFCCVEGCSA-N
Synonym r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, r-boc-3-methoxy-beta-phe-oh, boc-r-3-amino-3-3-methoxy-phenyl-propionic acid, boc-r-3-amino-3-3-methoxyphenyl propionic acid, r-3-boc-amino-3-3-methoxyphenyl propionic acid, 3r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, 3r-3-tert-butoxy carbonylamino-3-3-methoxyphenyl propanoic acid, pubchem24031, boc-d-?-phe 3-ome-oh, r-boc-3-methoxy-, a-phe-oh
PubChem CID 2761760
IUPAC Name (3R)-3-(3-methoxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)OC

Specifications

Melting Point 106°C to 108°C
Quantity 1 g
Optical Rotation +40° (c=1 in Ethanol)
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.