Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-3-(Boc-amino)pyrrolidine, 99%, ee 99%, Thermo Scientific™

Catalog No. AAL1969203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1969203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1969203 Supplier Thermo Scientific Chemicals Supplier No. L1969203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(+)-3-(Boc-amino)pyrrolidine is used as an intermediate in organic synthesis, as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122536-77-0
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00143191
InChI Key DQQJBEAXSOOCPG-SSDOTTSWSA-N
Synonym r-3-boc-amino pyrrolidine, r-tert-butyl pyrrolidin-3-ylcarbamate, r-+-3-boc-amino pyrrolidine, tert-butyl n-3r-pyrrolidin-3-yl carbamate, 3r-+-3-tert-butoxycarbonylamino pyrrolidine, r-3-n-boc-aminopyrrolidine, 3r-+-3-boc-amino pyrrolidine, r-3-boc-aminopyrrolidine, tert-butyl r-pyrrolidin-3-ylcarbamate, tert-butyl 3r-pyrrolidin-3-ylcarbamate
PubChem CID 1514397
IUPAC Name tert-butyl N-[(3R)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Melting Point 50°C to 54°C
UN Number UN3259
Beilstein 5377812
Solubility Information Soluble in water.
Quantity 1 g
Optical Rotation +21° (c=1 in Ethanol)
Formula Weight 186.25
Sensitivity Air sensitive
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-(+)-3-(Boc-amino)pyrrolidine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.