Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R)-(3-Chloro-phenyl)-[(9H-fluoren-9-ylmethoxycarbonylamino)]-acetic acid

Catalog No. CDS022475 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS022475 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS022475 Supplier Sigma Aldrich Supplier No. CDS022475100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(3-Chloro-phenyl)-[(9H-fluoren-9-ylmethoxycarbonylamino)]-acetic acid

Specifications

Molecular Formula C23H18ClNO4
Linear Formula C23H18ClNO4
MDL Number MFCD07371718
Quantity 1 g
Molecular Weight (g/mol) 407.85
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.