Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-3-hydroxybutanoic acid sodium | 13613-65-5 | MFCD00077788 | 99.9% | 126.09 | C4H7NaO3 | 10 G
SDP

Catalog No. 5000299917
Encompass_Preferred

(R)-3-Hydroxybutanoic acid sodium is the sodium salt of the endogenous metabolite R-3-hydroxybutyric acid supplied as a white to off-white solid for research applications. It has formula C4H7NaO3 and molecular weight 126.09. Reported purity is 99.90%. Manufacturer data include detailed solubility and storage guidance suitable for in vitro and in vivo use.

  • High stated purity (99.90%).
  • White to off-white solid suitable for analytical and formulation workflows.
  • High aqueous solubility: 100 mg/mL in water and PBS with ultrasonic assistance.
  • DMSO solubility up to 50 mg/mL with ultrasonic warming to 60°C.
  • Available in multiple pack sizes, including 10 g, for scaling experiments.
  • Recommended storage: sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99917 Supplier Medchemexpress LLC Supplier No. HYW01585110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-3-Hydroxybutanoic acid sodium is the sodium salt of the endogenous metabolite R-3-hydroxybutyric acid supplied as a white to off-white solid for research applications. It has formula C4H7NaO3 and molecular weight 126.09. Reported purity is 99.90%. Manufacturer data include detailed solubility and storage guidance suitable for in vitro and in vivo use.

  • High stated purity (99.90%).
  • White to off-white solid suitable for analytical and formulation workflows.
  • High aqueous solubility: 100 mg/mL in water and PBS with ultrasonic assistance.
  • DMSO solubility up to 50 mg/mL with ultrasonic warming to 60°C.
  • Available in multiple pack sizes, including 10 g, for scaling experiments.
  • Recommended storage: sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.