Learn More
AdipoGen R-3-Hydroxymyristic acid
SDP
Catalog No. 502560722
Encompass_Preferred
Chemical. CAS 28715-21-1. Formula C14H28O3. MW 244.4. Synthetic. Hydroxy fatty acid used to study its role in biological processes such as oxidative stress, inflammation and insulin resistance. Used in endotoxin and lipid A research, since it is one of two major components of bacterial Lipid A. Shows antifungal activity. Compound could be used as analytical reference in assaying food samples.
Catalog No. 50-256-0722 Supplier AdipoGen Supplier No. CDXH0114G001
Chemical. CAS 28715-21-1. Formula C14H28O3. MW 244.4. Synthetic. Hydroxy fatty acid used to study its role in biological processes such as oxidative stress, inflammation and insulin resistance. Used in endotoxin and lipid A research, since it is one of two major components of bacterial Lipid A. Shows antifungal activity. Compound could be used as analytical reference in assaying food samples.