Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R)-3-methyl-pyrrolidine hydrochloride

Catalog No. CDS006861 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS006861 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS006861 Supplier Sigma Aldrich Supplier No. CDS00686125MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Benzenesulfonyl isocyanate reacts with ethanol and phenol to yield normal urethan products.benzenesulfonyl isocyanate was used in synthesis of sterically hindered derivatives of phenols and alcohols
  • It was also used in the reaction of semi crystalline poly(ether ether ketone) films with carbonyl selective reagents to produce modified film samples

Specifications

Molecular Formula C5H12ClN
Linear Formula C5H12ClN
Quantity 10 g
MDL Number MFCD06796631
Molecular Weight (g/mol) 121.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.