Learn More
AdipoGen (R)-3-Hydroxybutyric acid, 625-72-9, MFCD00066257, 100 mg
SDP
Supplier:  AdipoGen AGCR13616M100
Encompass_Preferred
MF: C4H8O3, MW: 104.1, ≥98%, Appearance: White solid. Key metabolite of the ketolytic pathway generating acetyl-CoA. Energy carrier from adipocytes to peripheral tissues during fasting or exercise. Stereoselective product of D-3-hydroxybutyrate dehydreogenase. Clinically and physiologically significant stereoisomer.
Catalog No. 50-114-6578