Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-3-Piperidinamine dihydrochloride, 97%, Thermo Scientific™

Catalog No. AC436410010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC436410010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436410010 Supplier Thermo Scientific Chemicals Supplier No. 436410010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 334618-23-4
Molecular Formula C5H12N2·2ClH
Molecular Weight (g/mol) 173.09
InChI Key GGPNYXIOFZLNKW-ZJIMSODOSA-N
Synonym r-3-aminopiperidine dihydrochloride, r-piperidin-3-amine dihydrochloride, r---3-aminopiperidine dihydrochloride, r-3-piperidinamine dihydrochloride, 3r-piperidin-3-amine dihydrochloride, r-3-amino-piperidine dihydrochloride, r---3-aminopiperidine 2hcl, r-3-aminopiperidine 2hcl, r-3-amino piperidine dihcl
PubChem CID 16211333
IUPAC Name (3R)-piperidin-3-amine;dihydrochloride
SMILES C1CC(CNC1)N.Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 207.5°C to 211.5°C
Color Tan to White
Elemental Analysis 33.5 to 35.9% C, 15.5 to 16.8% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 1 g
Specific Rotation -1° to -3° (20°C, 589nm) (c=1, water)
Formula Weight 173.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-(-)-3-Piperidinamine dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED.
IF INHALED: Remove victim

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.