Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-3-Quinuclidinol, 99+%, Thermo Scientific Chemicals

Catalog No. AC293730025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC293730025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC293730025 Supplier Thermo Scientific Chemicals Supplier No. 293730025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25333-42-0
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00211251,MFCD13191732
InChI Key IVLICPVPXWEGCA-ZETCQYMHSA-N
Synonym r---3-quinuclidinol, r-quinuclidin-3-ol, r-3-quinuclidinol, 3r-quinuclidin-3-ol, 3-quinuclidinol l-form, unii-19xl022lm3, 3 r-quinuclidinol, 3r-1-azabicyclo 2.2.2 octan-3-ol, 3-r-quinuclidinol
PubChem CID 91404
IUPAC Name (3R)-1-azabicyclo[2.2.2]octan-3-ol
SMILES O[C@H]1CN2CCC1CC2

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 217.0°C to 224.0°C
Color Beige to White
Boiling Point 120.0°C (27.0 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Merck Index 15, 8199
Specific Rotation Condition − 44.50 (20.00°C c=3,1N HCl)
Specific Rotation − 44.50
Solubility Information Solubility in water: 1kg/l
Formula Weight 127.19
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-3-Quinuclidinol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 246-857-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.