Learn More
BUCHI Rotavapor R-300 Rotary Evaporators
GSA_VASDP
Catalog No. 05000461 Shop All BUCHI Products
Change view
Click to view available options
Condenser Type:
Cold Trap
Cold Trap for dry ice
Vertical
Vertical Coil for cold water or a chiller
Description:
Electronic lift
Manual lift
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Condenser Type Description
05-000-461 Cold Trap Manual lift
05-000-947 Cold Trap Electronic lift
05-000-977 Vertical Electronic lift
05-000-491 Vertical Manual lift
05-000-455 Cold Trap for dry ice Manual lift
05-000-485 Vertical Coil for cold water or a chiller Manual lift
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05-000-461 Supplier BUCHI Supplier No. 11R300152C102
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor R-300 to a fully integrated system where a central interface regulates each component.
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
Certifications
CE and CSA certified
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Description Manual lift
Voltage 100/120 V
Capacity (Metric) 5 L
Speed 10 to 280 rpm
Temperature Range (Metric) 20°C to 220°C
For Use With (Application) Distillation of Low Boiling-Point Solvents (for use with dry ice)
Condenser Type Cold Trap
Certifications CE / CSA
Type Rotary Evaporator
Frequency 50/60 Hz
Model R-300
Standard Taper Joint 24/40