Learn More
Strem, An Ascensus Company CAS# 364732-88-7. 100mg. (R)-(-)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane, min. 95% (R)-PHANEPHOS. MFCD03094575
SDP
Supplier:  Strem, An Ascensus Company 150425100MG
Encompass
CAS# 364732-88-7. 100mg. (R)-(-)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane, min. 95% (R)-PHANEPHOS. MFCD03094575. Molecular Weight: 576.65. Molecular Formula: C40H34P2. Color/form: white solid Strem# 15-0425. http://www.strem.com/catalog/v/15-0425/
Catalog No. 50-901-09858