Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ (R)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%

Catalog No. AA47278MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47278MD 250 mg
AA4727803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47278MD Supplier Thermo Scientific™ Supplier No. 47278MD
Add to Cart

Chemical Identifiers

Molecular Formula C{5}{0}H{5}{6}O{6}P{2}
Synonym (R)-(4,4',5,5',6,6'-Hexamethoxy[1,1'-biphenyl]-2,2'-diyl)bis[bis(3,5-dimethylphenyl]phosphane);

Specifications

Quantity 250 mg
Formula Weight 814.92
Percent Purity 97%
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.