Learn More
Medchemexpress LLC (R)-(4,4-difluoropiperidin-1-yl)(1-((4-(isopropylsulfonyl)phenyl)sulfonyl)piperidin-3-yl)methanone | 2749555-66-4 | 99.7% | C20H28F2N2O5S2 | 10MG
SDP
Catalog No. 5000258561
Encompass_Preferred
This research compound is a potent inhibitor of mitochondrial oxidative phosphorylation that targets Complex I and is supplied as a solid for laboratory research, with demonstrated low-nanomolar potency and oral activity in preclinical studies.
  • Highly potent complex I inhibitor (IC50 = 3.6 nM).
  • Impairs ATP generation in cells (IC50 = 11 nM).
  • Demonstrated oral bioavailability and in vivo efficacy in preclinical tumor models.
  • High chemical purity (99.7%) suitable for biochemical and cellular studies.
  • Supplied as a white to off-white solid with recommended low-temperature storage (powder: -20°C) for stability.
  • Molecular weight 478.57 g/mol to inform dosing and handling.
Catalog No. 50-002-58561 Supplier Medchemexpress LLC Supplier No. HY14431010MG
This research compound is a potent inhibitor of mitochondrial oxidative phosphorylation that targets Complex I and is supplied as a solid for laboratory research, with demonstrated low-nanomolar potency and oral activity in preclinical studies.
  • Highly potent complex I inhibitor (IC50 = 3.6 nM).
  • Impairs ATP generation in cells (IC50 = 11 nM).
  • Demonstrated oral bioavailability and in vivo efficacy in preclinical tumor models.
  • High chemical purity (99.7%) suitable for biochemical and cellular studies.
  • Supplied as a white to off-white solid with recommended low-temperature storage (powder: -20°C) for stability.
  • Molecular weight 478.57 g/mol to inform dosing and handling.