Learn More
Medchemexpress LLC (R)-4-Benzyl-2-oxazolidinone | 102029-44-7 | 177.20 | 500 G
SDP
Catalog No. 5000447727
Encompass_Preferred
(R)-4-Benzyl-2-oxazolidinone is an oxazolidinone-type auxiliary that enables highly stereoselective asymmetric alkylation reactions. It is used in the total synthesis of natural products and pharmaceutical preparations. In pharmaceutical research and development, it serves as a key precursor for the synthesis of aromatase inhibitors and can be used in studies related to estrogen-dependent breast cancer, where it serves as the core chiral parent nucleus of aromatase inhibitors. The inhibitory activity of its derivatives is significantly superior to that of (S)-4-Benzyl-2-oxazolidinone.
  • Enables highly stereoselective asymmetric alkylation reactions
  • Used in total synthesis of natural products
  • Used in pharmaceutical preparations
  • Key precursor for synthesis of aromatase inhibitors
  • Can be used in studies related to estrogen-dependent breast cancer
Catalog No. 50-004-47727 Supplier Medchemexpress LLC Supplier No. HYW007664500G
(R)-4-Benzyl-2-oxazolidinone is an oxazolidinone-type auxiliary that enables highly stereoselective asymmetric alkylation reactions. It is used in the total synthesis of natural products and pharmaceutical preparations. In pharmaceutical research and development, it serves as a key precursor for the synthesis of aromatase inhibitors and can be used in studies related to estrogen-dependent breast cancer, where it serves as the core chiral parent nucleus of aromatase inhibitors. The inhibitory activity of its derivatives is significantly superior to that of (S)-4-Benzyl-2-oxazolidinone.
  • Enables highly stereoselective asymmetric alkylation reactions
  • Used in total synthesis of natural products
  • Used in pharmaceutical preparations
  • Key precursor for synthesis of aromatase inhibitors
  • Can be used in studies related to estrogen-dependent breast cancer