Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane 97.0+%, TCI America™
SDP

Catalog No. B21361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21361G Supplier TCI America Supplier No. B21361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14347-83-2
Molecular Formula C13H18O3
Molecular Weight (g/mol) 222.284
MDL Number MFCD00066533
InChI Key DBFDSKSLTCMIPB-GFCCVEGCSA-N
PubChem CID 10944055
IUPAC Name (4R)-2,2-dimethyl-4-(phenylmethoxymethyl)-1,3-dioxolane
SMILES CC1(OCC(O1)COCC2=CC=CC=C2)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 222.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-4-Benzyloxymethyl-2,2-dimethyl-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.