Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 98%

Catalog No. AAH6295406
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6295406 5 g
AAH62954MD 250 mg
AAH6295403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6295406 Supplier Thermo Scientific Chemicals Supplier No. H6295406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 114360-56-4 | C24H28N2O6 | 440.496 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114360-56-4
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.496
MDL Number MFCD00798647
InChI Key LIWKOFAHRLBNMG-HXUWFJFHSA-N
Synonym fmoc-d-dab boc-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, n-a-fmoc-n-g-boc-d-2,4-diaminobutyric acid, 2r-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-gamma-t-butyloxycarbonyl-d-2,4-diaminobutyric acid, ambotzfaa1317, fmoc-4-boc-d-2,4-diaminobutyric acid, n-, a-fmoc-n-, a-boc-d-2,4-diaminobutyricacid, n-a-fmoc-n-gamma-boc-d-2,4-diaminobutyric acid, 2r-4-boc-amino-2-fmoc-amino-butanoic acid
PubChem CID 7021129
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Color White
Quantity 5 g
Formula Weight 440.49
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.