Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-4-Chloro-3-hydroxybutyronitrile 96.0+%, TCI America™
SDP

Catalog No. C27631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27631G 1 g
C27635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27631G Supplier TCI America Supplier No. C27631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84367-31-7
Molecular Formula C4H6ClNO
Molecular Weight (g/mol) 119.548
MDL Number MFCD00273362
InChI Key LHBPNZDUNCZWFL-SCSAIBSYSA-N
PubChem CID 641056
IUPAC Name (3R)-4-chloro-3-hydroxybutanenitrile
SMILES C(C#N)C(CCl)O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 119.55
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-4-Chloro-3-hydroxybutyronitrile
EINECSNumber (9)-0368&(9)-0460
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.