Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-4-Fluorostyrene oxide, ChiPros 98+%, ee 99%, Thermo Scientific™

Catalog No. AAL1958306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1958306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1958306 Supplier Thermo Scientific Chemicals Supplier No. L1958306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134356-73-3
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD03788740,MFCD00043994,MFCD03788740
InChI Key ICVNPQMUUHPPOK-UHFFFAOYNA-N
Synonym r-4-fluorophenyl oxirane, r-2-4-fluorophenyl oxirane, 2r-2-4-fluorophenyl oxirane, r-4-fluorostyrene oxide, r---4-fluorostyrene oxide, pubchem18166, r-4-fluorophenyl oxirane, technical gc
PubChem CID 7023123
IUPAC Name (2R)-2-(4-fluorophenyl)oxirane
SMILES FC1=CC=C(C=C1)C1CO1

Specifications

Boiling Point 92°C (14 mmHg)
Flash Point 77°C (171°F)
Quantity 5 g
UN Number UN1992
Beilstein 4350328
Optical Rotation −17° (c=1 in Chloroform)
Formula Weight 138.14
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-(-)-4-Fluorostyrene oxide
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H226-H351-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.