Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-4-Phenyl-2-butanol, 99+%, Thermo Scientific™

Catalog No. AAL18885MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18885MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL18885MD Supplier Thermo Scientific Chemicals Supplier No. L18885MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39516-03-5
Boiling Point 239°C
Flash Point >110°C (230°F)
Molecular Formula C10H14O
Quantity 250 mg
MDL Number MFCD03093081
Beilstein 5248382
Synonym r---4-phenyl-2-butanol, r-4-phenyl-2-butanol, r-4-phenylbutan-2-ol, unii-3r10q45i3f, 2r-4-phenylbutan-2-ol, --4-phenyl-2-butanol, 4-phenyl-2-butanol, r, r-alpha-methylbenzenepropanol, r-1-methyl-3-phenylpropanol, 4-phenyl-2-butanol, 2r
InChI Key GDWRKZLROIFUML-SECBINFHSA-N
Optical Rotation −14° (Neat)
SMILES CC(CCC1=CC=CC=C1)O
IUPAC Name (2R)-4-phenylbutan-2-ol
Molecular Weight (g/mol) 150.221
PubChem CID 641354
Formula Weight 150.22
Percent Purity ≥99%
Chemical Name or Material (R)-(-)-4-Phenyl-2-butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.