Learn More
Electron Microscopy Sciences MohrPro R-401 Dev/Activator/Cleaner 1 QT
SDP
Catalog No. 5024955
Encompass_Preferred
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.
Catalog No. 50-249-55 Supplier Electron Microscopy Sciences Supplier No. 7496503
All chemicals are concentrated . One quart (0.95L) of concentrate mixed with water will make one gallon of working solution.